missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MYCBP2 (aa 3364-3457) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84393 (PA5-84393. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes an E3 ubiquitin-protein ligase and member of the PHR (Phr1/MYCBP2, highwire and RPM-1) family of proteins. The encoded protein plays a role in axon guidance and synapse formation in the developing nervous system. In mammalian cells, this protein regulates the cAMP and mTOR signaling pathways, and may additionally regulate autophagy. Reduced expression of this gene has been observed in acute lymphoblastic leukemia patients and a mutation in this gene has been identified in patients with a rare inherited vision defect.
Specifications
Specifications
| Accession Number | O75592 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 23077 |
| Name | Human MYCBP2 (aa 3364-3457) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AU023734; AW107953; AW546647; C130061D10Rik; E3 ubiquitin-protein ligase MYCBP2; Highwire; KIAA0916; MYC binding protein 2; MYC binding protein 2, E3 ubiquitin protein ligase; Myc-binding protein 2; Mycbp2; Myc-bp2; PAM; pam, highwire, rpm 1; pam/highwire/rpm-1 protein; Phr; Phr1; probable E3 ubiquitin-protein ligase MYCBP2; protein associated with Myc; Protein Magellan; R75243 |
| Common Name | MYCBP2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction