missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MYL6B (aa 29-71) Control Fragment Recombinant Protein

Product Code. 30180577
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30180577

Brand: Invitrogen™ RP97888

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58579 (PA5-58579. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in both slow-twitch skeletal muscle and in nonmuscle tissue.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14649
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 140465
Name Human MYL6B (aa 29-71) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5730437E04Rik; BC037527; MLC1SA; MYL6B; myosin alkali light chain 1 slow a; myosin light chain 1 slow a; myosin light chain 1 slow-twitch muscle A isoform; myosin light chain 1, slow-twitch muscle A isoform; myosin light chain 6 B; myosin, light chain 6 B, alkali, smooth muscle and non-muscle; myosin, light polypeptide 6 B; myosin, light polypeptide 6 B, alkali, smooth muscle and non-muscle; sm; smooth muscle and non-muscle myosin alkali light chain 6 B; smooth muscle and nonmuscle myosin light chain alkali 6 B
Common Name MYL6B
Gene Symbol Myl6b
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence APPAKTKAEPAVPQAPQKTQEPPVDLSKVVIEFNKDQLEEFKE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.