missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Myocilin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF2537-SP
This item is not returnable.
View return policy
Description
Myocilin Polyclonal specifically detects Myocilin in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| Myocilin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GLC1A, GLC1Amyocilin, GPOA, JOAG, JOAG1, MYOC, myocilin, trabecular meshwork inducible glucocorticoid response, TIGR, TIGRmutated trabecular meshwork-induced glucocorticoid response protein, Trabecular meshwork-induced glucocorticoid response protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4653 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 50 ug/mL | |
| Q99972 | |
| MYOC | |
| Mouse myeloma cell line NS0-derived recombinant human Myocilin Arg33-Met504 Accession # Q99972 | |
| 25 μg | |
| Primary | |
| Detects human Myocilin in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction