Learn More
Human Myoglobin Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB97202-SP
 View more versions of this product
Product Code. 18151841
  • € 196.00 / 25µg
Description
Myoglobin Monoclonal specifically detects Myoglobin in Human samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Myoglobin
Monoclonal
Unconjugated
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
MB, MGC13548, myoglobin, PVALB
Purified human cardiac myoglobin antigen from human heart Accession # P02144
25 μg
4151
Reconstitute at 0.5mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a
ELISA, Immunohistochemistry, Western Blot
974145
Western Blot 0.05μg/mL, Simple Western 10μg/mL, ELISA, Immunohistochemistry 1-25μg/mL
P02144
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human Myoglobin in direct ELISAs. In sandwich immunoassays, the antibody pair is specific for human Myoglobin.
Human
Supernatant