missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MYRIP (aa 328-464) Control Fragment Recombinant Protein

Product Code. 30206116
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30206116

Brand: Invitrogen™ RP96787

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82250 (PA5-82250. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

MYRIP is a protein coding gene. Rab effector protein involved in melanosome transport. Serves as link between melanosome-bound RAB27A and the motor proteins MYO5A and MYO7A. May link RAB27A-containing vesicles to actin filaments. Functions as a protein kinase A-anchoring protein (AKAP). May act as a scaffolding protein that links PKA to components of the exocytosis machinery, thus facilitating exocytosis, including insulin release.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NFW9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 25924
Name Human MYRIP (aa 328-464) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A230081N12Rik; DKFZp586F1018; exophilin; exophilin 8; exophilin-8; FLJ44025; MGC130034; MGC130035; myosin VIIA and Rab interacting protein; myosin-VIIa- and Rab-interacting protein; MYRIP; OTTHUMP00000209206; Rab effector MyRIP; Slac2c; slaC2-c; slp homolog lacking C2 domains c; Slp homologue lacking C2 domains; synaptotagmin-like protein homologue lacking C2 domains-c; synaptotagmin-like protein lacking C2 domains C
Common Name MYRIP
Gene Symbol MYRIP
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SALPSWKSVDRLDETNLAPVLQSPDGNWVALKDGAPPPTRLLAKPKSGTFQALEVASSVASAYDEMGSDSEEDFDWSEALSKLCPRSRALPRNPQPQPTQAQSSDQGPIAASPSSALSPNPEAMCSDSETSSAGSSR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.