missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NBEAL1 (aa 2541-2640) Control Fragment Recombinant Protein

Product Code. 30180613
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
30180613 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 30180613

Brand: Invitrogen™ RP99648

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61974 (PA5-61974. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

NBEAL1 (Neurobeachin Like 1) is a Protein Coding gene. Diseases associated with NBEAL1 include Ovary Serous Adenocarcinoma. Gene Ontology (GO) annotations related to this gene include binding. An important paralog of this gene is NBEAL2.
TRUSTED_SUSTAINABILITY

Specifikationer

Accession Number Q6ZS30
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 65065
Name Human NBEAL1 (aa 2541-2640) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A530083I02Rik; ALS2CR16; ALS2CR17; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 16; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 17; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 16 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 17 protein; beach; NBEAL1; neurobeachin like 1; neurobeachin-like 1; neurobeachin-like protein 1
Common Name NBEAL1
Gene Symbol NBEAL1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TIPNLAISWEGHIVVYSSTEEKTTLKDKNALHLFSINGKYLGSQILKEQVSDICIIGEHIVTGSIQGFLSIRDLHSLNLSINPLAMRLPIHCVCVTKEYS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.