missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Nestin Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC1259S-100UG
This item is not returnable.
View return policy
Description
Nestin Monoclonal specifically detects Nestin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Nestin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| FLJ21841, NES, nestin | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Nestin. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 196908 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| NES | |
| NS0 mouse myeloma cell line transfected with human Nestin | |
| RUO | |
| 10763 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction