missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Netrin-G1a Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB10019G-100UG
This item is not returnable.
View return policy
Description
Netrin-G1a Monoclonal specifically detects Netrin-G1a in Human samples. It is validated for Flow Cytometry.
Specifications
| Netrin-G1a | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 22854 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 1010035 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9Y212 | |
| Human embryonic kidney cell, HEK293-derived human Netrin-G1a, His29-Gly503, Accession # Q9Y212 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction