missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neurogenin-3 Biotinylated Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems BAF3444
This item is not returnable.
View return policy
Description
Neurogenin-3 Polyclonal specifically detects Neurogenin-3 in Human samples. It is validated for Western Blot.
Specifications
| Neurogenin-3 | |
| Polyclonal | |
| Biotin | |
| Q9Y4Z2 | |
| NEUROG3 | |
| E. coli-derived recombinant human Neurogenin-3 Met1-Leu214 Accession # Q9Y4Z2 | |
| 50 μg | |
| Primary | |
| Detects human Neurogenin-3 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human Neurogenin-1 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Atoh5, BHLHA7, bHLHa7NGN-3, Class A basic helix-loop-helix protein 7, Math4B, NEUROG3, neurogenin 3, neurogenin-3, Neurogenin3, NGN3, Protein atonal homolog 5, Relax | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 50674 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction