missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neuromedin BR/NMBR Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC4728N-100UG
This item is not returnable.
View return policy
Description
Neuromedin BR/NMBR Monoclonal specifically detects Neuromedin BR/NMBR in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Neuromedin BR/NMBR | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| AAB27330 | |
| NMBR | |
| NS0 mouse myeloma cell line transfected with human NMBR Met1-Met390 Accession # AAB27330 | |
| RUO | |
| 4829 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 466505 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Neuromedin B R, neuromedin B receptor, neuromedin-B receptor, Neuromedin-B-preferring bombesin receptor, NMBR, NMB-R | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Neuromedin B R/NMBR. Stains human Neuromedin B R/NMBR transfectants but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction