missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NF-H Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3108
This item is not returnable.
View return policy
Description
NF-H Polyclonal specifically detects NF-H in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| NF-H | |
| Polyclonal | |
| Unconjugated | |
| P12036 | |
| NEFH | |
| E. coli-derived recombinant human NF-H Met1-Ala380 Accession # P12036 | |
| 100 μg | |
| Primary | |
| Detects human NF-H in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) NF-M and rhNF-L is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| 200 kDa neurofilament protein, KIAA0845, NEFH, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4744 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction