missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NHA2/SLC9B2 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB8404S-100UG
This item is not returnable.
View return policy
Description
NHA2/SLC9B2/NHEDC2 Monoclonal specifically detects NHA2/SLC9B2/NHEDC2 in Human samples. It is validated for Flow Cytometry.
Specifications
| NHA2/SLC9B2/NHEDC2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_849155 | |
| SLC9B2 | |
| NS0 mouse myeloma cell line transfected with human NHA2/SLC9B2 Met1-Val537 Accession # NP_849155 | |
| RUO | |
| 133308 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 896151 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| FLJ23984, Mitochondrial Na(+)/H(+) exchanger NHA2, mitochondrial sodium/hydrogen exchanger NHA2, Na(+)/H(+) exchanger-like domain-containing protein 2, Na+/H+ exchanger domain containing 2, NHA2NHE10, NHE domain-containing protein 2, NHE10, NHEDC2, SLC9B2, Sodium/hydrogen exchanger-like domain-containing protein 2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects HEK293 cells transfected with human NHA2/SLC9B2 by Flow Cytometry. Does not detect untransfected or irrelevant transfected HEK293 cells. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction