missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Nidogen-1/Entactin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Brand: R&D Systems AF2570
This item is not returnable.
View return policy
Description
Nidogen-1/Entactin Polyclonal specifically detects Nidogen-1/Entactin in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Nidogen-1/Entactin | |
| Polyclonal | |
| Unconjugated | |
| AAH45606 | |
| NID1 | |
| Mouse myeloma cell line NS0-derived recombinant human Nidogen-1/Entactin Leu29-Lys1114 (Gln1113Arg) Accession # AAH45606 | |
| 100 μg | |
| Primary | |
| Detects human Nidogen-1/Entactin in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| enactin, Entactin, Entactin-1, NID1, NID-1, NIDentactin, nidogen (enactin), nidogen 1, nidogen-1, Nidogen1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4811 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction