missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human NKG2A/CD159a (NP_002250) VersaClone cDNA

Product Code. 18714909
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18714909 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18714909 Supplier R&D Systems™ Supplier No. RDC2583
  • ONLINE EXCLUSIVE PRICE
    € 328.00
    € 309.75 / 10µg
    Save € 18.25
    5% Off
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

  • KLRC1 killer cell lectin like receptor C1 [ Homo sapiens (human) ]
  • Also known as: NKG2; NKG2A; CD159A
  • NKG2A and NKG2C are type II transmembrane proteins with extracellular C-type lectin domains.
  • These proteins heterodimerize with CD94 to form complexes that recognize HLA-E as ligand.
  • The NKG2A/CD94 complex delivers an inhibitory signal resulting from ITIMs within the NKG2A cytoplasmic domain.
  • NKG2C/CD94 provides an activating signal due to association with the ITAM-containing DAP12 adapter protein.
  • These complexes are expressed on NK and T cell subsets.
  • The NKG2A/CD159a/KLRC1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • For reasearch use only. Not for use in humans.
TRUSTED_SUSTAINABILITY

Specifications

Gene hKLRC1
Accession Number NP_002250
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 715 bp
Gene ID (Entrez) 3821
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.