missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKG2D/CD314 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB139V-100UG
This item is not returnable.
View return policy
Description
NKG2D/CD314 Monoclonal specifically detects NKG2D/CD314 in Human samples. It is validated for Flow Cytometry.
Specifications
| NKG2D/CD314 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, KLRK1, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 22914 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 149810 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| KLRK1 | |
| BaF3 mouse pro-B cell line transfected with human NKG2D/CD314 | |
| 100 μg | |
| Primary | |
| Detects human NKG2D/CD314 in Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction