missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKG2D/CD314 Antibody, R&D Systems™

Mouse Monoclonal Antibody
€ 218.00 - € 569.00
Specifications
| Antigen | NKG2D/CD314 |
|---|---|
| Clone | 1046002 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18745785
![]() |
R&D Systems
MAB1392-100 |
100 μg |
€ 569.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18765745
![]() |
R&D Systems
MAB1392-SP |
25 μg |
€ 218.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKG2D/CD314 Monoclonal antibody specifically detects NKG2D/CD314 in Human samples. It is validated for Flow CytometrySpecifications
| NKG2D/CD314 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, KLRK1, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence | |
| Mouse myeloma cell line NS0-derived recombinant human NKG2D/CD314, Phe78-Val216, Accession # P26718 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1046002 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P26718 | |
| 22914 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title