missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKX2.5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB24441-SP
This item is not returnable.
View return policy
Description
NKX2.5 Monoclonal specifically detects NKX2.5 in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| NKX2.5 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| P52952 | |
| NKX2-5 | |
| E. coli-derived recombinant human NKX2.5 Gln24-Arg137 Accession # P52952 | |
| 25 μg | |
| Primary | |
| Detects endogenous human NKX2.5 in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 259415 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cardiac-specific homeo box, Cardiac-specific homeobox, cardiac-specific homeobox 1, CHNG5, CSX1, CSXFLJ97195, FLJ52202, FLJ99536, Homeobox protein CSX, Homeobox protein NK-2 homolog E, homeobox protein Nkx-2.5, NK2 transcription factor related, locus 5 (Drosophila), NKX2.5FLJ97166, NKX2-5, NKX2E, NKX2EFLJ97197, NKX4-1, tinman paralog | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1482 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction