missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Nodal Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3218-SP
This item is not returnable.
View return policy
Description
Nodal Monoclonal specifically detects Nodal in Human samples. It is validated for Western Blot.
Specifications
| Nodal | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q96S42 | |
| NODAL | |
| E. coli-derived recombinant human Nodal His238-Leu347 Accession # Q96S42 | |
| 25 μg | |
| Primary | |
| Detects human Nodal in ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant mouse Nodal is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 784410 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP-16, MGC138230, nodal homolog, nodal homolog (mouse), nodal, mouse, homolog | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4838 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction