missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Notch-1 Intracellular Domain Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems BAF3647
This item is not returnable.
View return policy
Description
Notch-1 Polyclonal specifically detects Notch-1 in Human samples. It is validated for Western Blot.
Specifications
| Notch-1 | |
| Polyclonal | |
| Biotin | |
| AAG33848 | |
| NOTCH1 | |
| E. coli-derived recombinant human Notch-1 Gly2428-Lys2556 Accession # AAG33848 | |
| 50 μg | |
| Primary | |
| Detects human Notch-1 Intracellular Domain (ICD) in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) Notch-2 ICD, rhNotch-3 ICD, and rhNotch-4 ICD is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, Notch1, TAN1, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4851 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction