missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NOV/CCN3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF1640-SP
This item is not returnable.
View return policy
Description
NOV/CCN3 Polyclonal specifically detects NOV/CCN3 in Human samples. It is validated for Western Blot, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| NOV/CCN3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CCN family member 3, CCN3, CCN3IGF-binding protein 9, IBP-9, IGFBP-9, IGFBP9novH, Insulin-like growth factor-binding protein 9, nephroblastoma overexpressed gene, Nephroblastoma-overexpressed gene protein homolog, NOVH, protein NOV homolog | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4856 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| P48745 | |
| NOV | |
| Mouse myeloma cell line NS0-derived recombinant human NOV/CCN3 Thr32-Met357 Accession # P48745 | |
| 25 μg | |
| Primary | |
| Detects human NOV/CCN3 in ELISAs and Western blots. In sandwich ELISAs, approximately 7% cross-reactivity with recombinant mouse NOV/CCN3 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction