Learn More
R&D Systems™ Human NPRC/NPR3 (NP_001191304) VersaClone cDNA
Description
The Human NPRC/NPR3 (NP_001191304) VersaClone cDNA clone contains the complete ORF for Natriuretic Peptide Receptor C, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Natriuretic Peptide Receptor C insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Specifications
Specifications
| Gene | hNPR3 |
| Accession Number | NP_001191304 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl |
| Insert Type | 1639bp |
| pH | 8.5 |
| Gene ID (Entrez) | 4883 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |