missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NPY1R Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB6400S-100UG
This item is not returnable.
View return policy
Description
NPY1R Monoclonal specifically detects NPY1R in Human samples. It is validated for Flow Cytometry.
Specifications
| NPY1R | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P25929 | |
| NPY1R | |
| NS0 mouse myeloma cell line transfected with human NPY1R Met1-Ile384 Accession # P25929 | |
| RUO | |
| 4886 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 556153 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| neuropeptide Y receptor type 1, neuropeptide Y receptor Y1, NPY1-R, NPYR, NPYY1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human NPY1R in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction