Learn More
Human NrCAM Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2034
This item is not returnable.
View return policy
Description
NrCAM Monoclonal specifically detects NrCAM in Human samples. It is validated for Western Blot.
Specifications
| NrCAM | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q14CA1 | |
| NRCAM | |
| Mouse myeloma cell line NS0-derived recombinant human NrCAM isoform 3 Leu30-Asn600 (Ala526Pro) Accession # Q14CA1 | |
| 100 μg | |
| Primary | |
| Detects human NrCAM in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 297906 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| bravo, hBravo, KIAA0343Neuronal surface protein Bravo, MGC138845, MGC138846, neuronal cell adhesion molecule, Ng-CAM-related, NgCAM-related cell adhesion molecule, nr-CAM | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4897 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |