Learn More
Human NT-3 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems AF-267-NA
Product Code. 18082202
  • € 631.00 / 100µg
Description
NT-3 Polyclonal specifically detects NT-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NT-3
Polyclonal
Unconjugated
P20783
NTF3
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human NT-3 Tyr139-Thr257 Accession # P20783
100 μg
Primary
Detects human NT-3 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) NT-4 and rhBDNF is observed.
Human
IgG
Western Blot, Immunohistochemistry, Neutralization
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.1-0.4 ug/mL
HDNF, MGC129711, Nerve growth factor 2, Neurotrophic factor, neurotrophin 3, neurotrophin-3, NGF2, NGF-2, NT3, NT-3, NTF3
Goat
Affinity Purified
RUO
4908
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.