missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NT-4 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF268
This item is not returnable.
View return policy
Description
NT-4 Polyclonal specifically detects NT-4 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| NT-4 | |
| Polyclonal | |
| Biotin | |
| P34130 | |
| NTF4 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human NT-4 Gly81-Ala210 Accession # P34130 | |
| 50 μg | |
| Primary | |
| Detects human NT-4 in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human (rh) CNTF, rhNeurturin, rhNT-3, rhBDNF, rhGDNF, and rh beta-NGF is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| GLC10, GLC1O, neurotrophic factor 4, neurotrophic factor 5, neurotrophin 4, neurotrophin 5 (neurotrophin 4/5), neurotrophin-4, neurotrophin-5, Neutrophic factor 4, NT4, NT-4, NT-4/5, NT-5, NTF4, NTF5, NTF5NT5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4909 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction