missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human nuclei Mouse anti-Human, Clone: 235-1 (PE), Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14206.500uL
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against nuclei in human cells.
- High specificity
- High reactivity
Specifications
| Human nuclei | |
| Monoclonal | |
| PE | |
| Flow Cytometry (5 uL/test/million cells in 0.1 mL or 5 uL/100 uL of whole blood) Immunofluorescence (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Protein G purification | |
| RUO | |
| This antibody does not react with nuclei of mouse, rat and chicken. | |
| Store at 4°C. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence | |
| 235-1 | |
| Mouse monoclonal antibody raised against nuclei in human cells. | |
| In PBS (0.05% BSA, 0.05% sodium azide). | |
| Nuclei of human myeloid leukemia biopsy cells. | |
| 500 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction