Learn More
R&D Systems™ Human OAT1 (NP_004781) VersaClone cDNA
Product Code. 18724248
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18724248 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18724248 Supplier R&D Systems™ Supplier No. RDC2587
  • ONLINE EXCLUSIVE PRICE
    € 328.00
    € 309.75 / 10µg
    Save € 18.25
    5% Off
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.
  • The Human OAT1 (NP_004781) VersaClone cDNA clone contains the complete ORF for SLC22A6, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The SLC22A6 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY
Specifications
Gene hOAT1
Accession Number NP_004781
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gene ID (Entrez) 9356
Content And Storage Store the unopened product at -20 to -80°C
Species Human
Quantity 10 μg