missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ORAI2 (aa 171-194) Control Fragment Recombinant Protein

Product Code. 30205300
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205300

Brand: Invitrogen™ RP103614

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (67%), Rat (67%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64250 (PA5-64250. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Antigen stimulation of immune cells triggers Ca++ entry through Ca++ release-activated Ca++ (CRAC) channels. ORAI2 is one of two mammalian homologs to ORAI1, a recently identified four-transmembrane spanning protein that is an essential component of CRAC. Like ORAI1, ORAI2 has been shown to function as a highly selective Ca++ plasma membrane channel that is gated through interactions with STIM1, the store-activated endoplasmic reticulum Ca++ sensor, although at a lesser efficacy than ORAI1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96SN7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 80228
Name Human ORAI2 (aa 171-194) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A730041O15Rik; C7orf19; calcium release-activated calcium channel 2; calcium release-activated calcium modulator 2 protein; CAP-binding protein complex interacting protein 2; CAP-binding protein complex-interacting protein 2; CBCIP2; H_NH0514P08.8; MEM142B; ORAI calcium release-activated calcium modulator 2; ORAI2; PP1729; Protein orai-2; putative protein ORAI2-2; RGD1310213; TMEM142B; Transmembrane protein 142 B
Common Name ORAI2
Gene Symbol ORAI2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IKFLPVDARRQPGPPPGPGSHTGW
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.