missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Orexin R2/HCRTR2 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC5246U-100UG
This item is not returnable.
View return policy
Description
Orexin R2/HCRTR2 Monoclonal specifically detects Orexin R2/HCRTR2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Orexin R2/HCRTR2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| AAC39602 | |
| HCRTR2 | |
| NS0 mouse myeloma cell line transfected with human Orexin R2/HCRTR2 Met1-Trp444 Accession # AAC39602 | |
| RUO | |
| 3062 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 456738 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| HCRTR2, hypocretin (orexin) receptor 2, Hypocretin receptor type 2, Orexin Receptor 2, orexin receptor type 2, OrexinR2, Ox-2-R, Ox2-R, OX2R | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Orexin R2/HCRTR2. Stains human Orexin R2/HCRTR2 transfectants but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction