missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Orexin R2/HCRTR2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5246-SP
This item is not returnable.
View return policy
Description
Orexin R2/HCRTR2 Monoclonal specifically detects Orexin R2/HCRTR2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| Orexin R2/HCRTR2 | |
| Monoclonal | |
| LYOPH | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| AAC39602 | |
| HCRTR2 | |
| NS0 mouse myeloma cell line transfected with human Orexin R2/HCRTR2 Met1-Trp444 Accession # AAC39602 | |
| 25 μg | |
| Primary | |
| Detects human Orexin R2/HCRTR2. Stains human Orexin R2/HCRTR2 transfectants but not irrelevant transfectants. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 456738 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HCRTR2, hypocretin (orexin) receptor 2, Hypocretin receptor type 2, Orexin Receptor 2, orexin receptor type 2, OrexinR2, Ox-2-R, Ox2-R, OX2R | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3062 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction