missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human OX40 Ligand/TNFSF4 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB10541N-100UG
This item is not returnable.
View return policy
Description
OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Human samples. It is validated for Flow Cytometry.
Specifications
| OX40 Ligand/TNFSF4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P23510 | |
| TNFSF4 | |
| Mouse myeloma cell line NS0-derived recombinant human OX40 Ligand Gln51-Leu183 Accession # P23510 | |
| RUO | |
| 7292 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 159403 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, TNFSF4, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human OX40 Ligand in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) APRIL, rhBAFF, rhCD27 Ligand, recombinant mouse (rm) CD27 Ligand, rhCD30 Ligand, rmCD30 Ligand, rhCD40 Ligand, rmEDA, rhFas Ligand, rmFas Ligand, rhGITR Ligand, recombinant cotton rat TNF-alpha , rhTNF-alpha , rmTNF-alpha , recombinant porcine TNF-alpha , recombinant rat TNF-alpha , rhTRAIL, rhTRANCE, rmTRANCE, or rhVEGF is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction