missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human OX40/TNFRSF4 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB10543R-100UG
This item is not returnable.
View return policy
Description
OX40/TNFRSF4 Monoclonal specifically detects OX40/TNFRSF4 in Human samples. It is validated for Flow Cytometry.
Specifications
| OX40/TNFRSF4 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| ACT-135, ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, Ly-70, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, TNFRSF4, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, Txgp1, TXGP1L, TXGP1LOX40 antigen | |
| Mouse myeloma cell line NS0-derived recombinant human OX40/TNFRSF4, Leu29-Ala216, Accession No. P43489 | |
| 100 μg | |
| 7293.0 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 977974 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| P43489 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human OX40/TNFRSF4 in direct ELISAs | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction