missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human P2X1/P2RX1 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB10533S-100UG
This item is not returnable.
View return policy
Description
P2X1/P2RX1 Monoclonal antibody specifically detects P2X1/P2RX1 in Human samples. It is validated for Flow Cytometry
Specifications
| P2X1/P2RX1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| ATP receptor, P2RX1, P2X purinoceptor 1, P2X receptor, subunit 1, P2X1P2X1 receptor, P2XMR, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 1 | |
| Human embryonic kidney cell, HEK293-derived transfected with human P2X1/P2RX1, Accession # P51575 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 551820 | |
| Flow Cytometry 0.25-1 ug/106 cells | |
| P51575 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5023 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction