missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human p300 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF3789
This item is not returnable.
View return policy
Description
KAT3B/p300 Polyclonal specifically detects KAT3B/p300 in Human samples. It is validated for Western Blot.
Specifications
| KAT3B/p300 | |
| Polyclonal | |
| Biotin | |
| Q09472 | |
| EP300 | |
| E. coli-derived recombinant human p300 His2283-His2414 Accession # Q09472 | |
| 50 μg | |
| Primary | |
| Detects human p300 in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, EP300, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2033 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction