missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human p53 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC13551V-100UG
This item is not returnable.
View return policy
Description
p53 Monoclonal specifically detects p53 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| p53 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P04637 | |
| TP53 | |
| E. coli-derived recombinant human p53 Asp7-Asp393 Accession # P04637 | |
| RUO | |
| 7157 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 184727 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Antigen NY-CO-13, BCC7, FLJ92943, LFS1, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, TP53, transformation-related protein 53, TRP53, tumor protein p53, Tumor suppressor p53 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human p53. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction