missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pannexin-2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB7088
This item is not returnable.
View return policy
Description
Pannexin-2 Monoclonal specifically detects Pannexin-2 in Human samples. It is validated for Western Blot.
Specifications
| Pannexin-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| Q96RD6 | |
| PANX2 | |
| E. coli-derived recombinant human Pannexin-2 Arg316-Lys456 Accession # Q96RD6 | |
| 100 μg | |
| Primary | |
| Detects human Pannexin-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human (rh) Pannexin-1 and no cross-reactivity with rhPannexin-3 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 719406 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| hPANX2, MGC119432, pannexin 2, pannexin-2, Pannexin2, PANX2, PX2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 56666 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction