missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pax6 PE-conjugated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems IC8150P-100
This item is not returnable.
View return policy
Description
Pax6 Polyclonal specifically detects Pax6 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Pax6 | |
| Polyclonal | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| P26367 | |
| Sheep | |
| Affinity Purified | |
| Primary | |
| Detects human Pax6 in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Pax1, rhPax2, rhPax3, rhPax4, rhPax5, and rhPax7 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6 | |
| E. coli-derived recombinant human Pax6 Met1-Arg272 Accession # P26367 | |
| 100 Tests | |
| 5080 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction