missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PCID2 (aa 1-71) Control Fragment Recombinant Protein

Product Code. 30208126
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30208126

Brand: Invitrogen™ RP104217

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64400 (PA5-64400. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Required for B-cell survival through the regulation of the expression of cell-cycle checkpoint MAD2L1 protein during B cell differentiation. Component of the TREX-2 complex (transcription and export complex 2), composed of at least ENY2, GANP, PCID2, DSS1, and either centrin CETN2 or CETN3 (PubMed:22307388). The TREX-2 complex functions in docking export-competent ribonucleoprotein particles (mRNPs) to the nuclear entrance of the nuclear pore complex (nuclear basket). TREX-2 participates in mRNA export and accurate chromatin positioning in the nucleus by tethering genes to the nuclear periphery. Binds and stabilizes BRCA2 and is thus involved in the control of R-loop-associated DNA damage and transcription-associated genomic instability. R-loop accumulation does not increase in PCID2-depleted cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5JVF3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55795
Name Human PCID2 (aa 1-71) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A730042J05Rik; CSN12-like protein; F10; fa11f03; HT004; hypothetical protein LOC617943; ik:tdsubc_1e10; im:6909928; PCI domain containing 2; PCI domain-containing protein 2; Pcid2; RGD1307041; RP11-98F14.6; tdsubc_1e10; wu:fa11f03; xx:tdsubc_1e10; zgc:101845
Common Name PCID2
Gene Symbol PCID2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MAHITINQYLQQVYEAIDSRDGASCAELVSFKHPHVANPRLQMASPEEKCQQVLEPPYDEMFAAHLRCTYA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.