missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PD-1 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB10863V-100UG
This item is not returnable.
View return policy
Description
PD-1 Monoclonal specifically detects PD-1 in Human samples. It is validated for Flow Cytometry.
Specifications
| PD-1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide | |
| CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| Mouse myeloma cell line, NS0-derived human PD-1, Met1-Gln167, Accession No. Q15116 | |
| Primary | |
| Detects human PD-1 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. |
| Flow Cytometry | |
| 2335A | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| Q15116 | |
| Rabbit | |
| 100 μg | |
| 5133 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction