missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PD-1 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB10861R-100UG
This item is not returnable.
View return policy
Description
PD-1 Monoclonal specifically detects PD-1 in Human samples. It is validated for Flow Cytometry.
Specifications
| PD-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q15116 | |
| PDCD1 | |
| Mouse myeloma cell line NS0-derived recombinant human PD-1 Met1-Gln167 Accession # Q15116 | |
| 100 μg | |
| Primary | |
| Detects human PD-1 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| 913429 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 5133 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction