missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PD-1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
€ 201.00 - € 462.00
Specifications
| Antigen | PD-1 |
|---|---|
| Clone | 1015829 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction |
| Applications | Flow Cytometry, Blocking Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18172721
|
R&D Systems
MAB10868-100 |
100 μg |
€ 462.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18130441
|
R&D Systems
MAB10868-SP |
25 μg |
€ 201.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Flow Cytometry, Blocking AssaySpecifications
| PD-1 | |
| Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| Human embryonic kidney cell HEK293-derived human PD-1, Leu25-Thr168, Accession # Q15116 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1015829 | |
| Flow Cytometry, Blocking Assay | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q15116 | |
| 5133 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title