Learn More
Human PD-L2/B7-DC Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB11351-SP
 View more versions of this product
Product Code. 18110931
  • € 207.00 / 25µg
Description
PD-L2/B7-DC/PDCD1LG2 Monoclonal antibody specifically detects PD-L2/B7-DC/PDCD1LG2 in Human samples. It is validated for Blocking Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PD-L2/B7-DC/PDCD1LG2
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, Butyrophilin-like Protein, CD273, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDCD1LG2, PDL2, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2
Human embryonic kidney cell, HEK293-derived recombinant human PD-L2/B7-DC, Accession # Q9BQ51
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
IgG1
Blocking Assay
1054620
Blockade of Receptor-ligand Interaction
Q9BQ51
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
80380