missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PEAR1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF4527
This item is not returnable.
View return policy
Description
PEAR1 Polyclonal specifically detects PEAR1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| PEAR1 | |
| Polyclonal | |
| Unconjugated | |
| Q5VY43 | |
| PEAR1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human PEAR1 Leu21-Ser754 Accession # Q5VY43 | |
| 100 μg | |
| Primary | |
| Detects human PEAR1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| JEDI, platelet endothelial aggregation receptor 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 375033 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction