missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pepsinogen A Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB7525
This item is not returnable.
View return policy
Description
Pepsinogen A Monoclonal specifically detects Pepsinogen A in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Pepsinogen A | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P00790 | |
| PGA4 | |
| Mouse myeloma cell line NS0-derived recombinant human Pepsinogen A4/PGA4 Ile16-Ala388 Accession # P00790 | |
| 100 μg | |
| Primary | |
| Detects human Pepsinogen A4/PGA4 in direct ELISAs. Human PGA3 and PGA5 reactivity has not been demonstrated but is likely due to their high homology with human PGA4. In direct ELISAs, no cross-reactivity with recombinant human (rh) Pepsinogen C, rhCathepsin D, or rhCathepsin E is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 762015 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.23, EC 3.4.23.1, FLJ58952, FLJ77962, pepsin A, pepsinogen 4, group I (pepsinogen A), pepsinogen A4, PGA3, PGA5 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 643847 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction