missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Perforin Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC103853R-100UG
This item is not returnable.
View return policy
Description
Perforin Monoclonal antibody specifically detects Perforin in Human samples. It is validated for Flow Cytometry
Specifications
| Perforin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFP, PFPcytolysin, PRF1 | |
| Chinese Hamster Ovary cell line CHO-derived human Perforin, Pro22-Trp555, Accession # P14222 | |
| 100 μg | |
| Primary | |
| Reconstitution default not known | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 1031751 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P14222 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5551 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction