missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSLP Antibody (55N2D4), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27325AF350
This item is not returnable.
View return policy
Description
TSLP Monoclonal specifically detects TSLP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TSLP | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TSLP | |
| amino acids (27-158) MDYDFTNCDFEkIkAAYLSTISkDLITYMSGTkSTEFNNTVSCSNRPHCLTEIQSLTFNP∼TAGCASLAkEMFAMkTkAALAIWCPGYSETQINATQAMkkRRkRkVTTNkCLEQVSQLQG∼LWRRFNRPLLkQQL | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 55N2D4 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| thymic stromal lymphopoietin | |
| Mouse | |
| Protein G purified | |
| Biologically Active Proteins, Innate Immunity | |
| 85480.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction