missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Periostin/OSF-2 DuoSet ELISA
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
The Human Periostin/OSF-2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Periostin/OSF-2.
Specifications
Specifications
| Assay Range | 62.50 - 4000pg/mL |
| Conjugate | Biotin |
| Immunoassay Kit Format | 96-well Strip Plate |
| Product Type | Solid Phase Sandwich ELISA |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 10631 (Human); 50706 (Mouse); 361945 (Rat) |
| Kit Contents | Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction