missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-CROCC (S1476) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
€ 234.00 - € 547.00
Specifications
| Antigen | CROCC |
|---|---|
| Clone | 2734A |
| Dilution | Western Blot 1 μg/mL, Immunocytochemistry 3-15 μg/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18704354
|
R&D Systems
MAB11335-SP |
25 μg |
€ 234.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18794384
![]() |
R&D Systems
MAB11335-100 |
100 μg |
€ 547.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CROCC Monoclonal antibody specifically detects CROCC in Human samples. It is validated for Western Blot, ImmunocytochemistrySpecifications
| CROCC | |
| Western Blot 1 μg/mL, Immunocytochemistry 3-15 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CROCC ciliary rootlet coiled-coil, rootletin, CROCC1, KIAA0445, ROLT, TAX1BP2 | |
| Phosphopeptide containing the human CROCC S site 1476, Accession # Q5TZA2 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 2734A | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q5TZA2 | |
| 9696 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title