missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-GCN2 (T899) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF7605-SP
This item is not returnable.
View return policy
Description
GCN2 Polyclonal specifically detects GCN2 in Human samples. It is validated for Western Blot.
Specifications
| GCN2 (p thr899) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EIF2AK4 | |
| Phosphopeptide containing human GCN2 T899 site | |
| 25 μg | |
| Primary | |
| Detects human GCN2 when phosphorylated at T899 in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| EIF2AK4, eukaryotic translation initiation factor 2 alpha kinase 4, eukaryotic translation initiation factor 2-alpha kinase 4, GCN2GCN2 eIF2alpha kinase, GCN2-like protein, KIAA1338EC 2.7.11.1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 440275 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction