missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-LAT (Y161) Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB6334
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
LAT Monoclonal specifically detects LAT in Human samples. It is validated for Western Blot.
Tekniske data
| LAT (p tyr161) | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| O43561 | |
| LAT | |
| Phosphopeptide containing the human LAT Y161 site Accession # O43561 | |
| 100 μg | |
| Primary | |
| Detects human Phospho-LAT (Y161) in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 627945 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27040 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion